2-Dibenzofuranamine 95%

Supplier: Ambeed
Warning

A334844-100MG A334844-1G A334844-250MG
77263-662EA 8 USD
77263-662 77263-664 77263-666
2-Dibenzofuranamine 95%
2-Dibenzofuranamine

Precaución: for Research Use Only

Formula: C₁₂H₉NO
MDL Number: MFCD00033416
CAS Number: 3693-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR