3,3-Dimethylmorpholine 97%

Supplier: Ambeed
Warning

A334648-100MG A334648-1G A334648-5G
77277-866EA 10 USD
77277-866 77277-868 77277-870
3,3-Dimethylmorpholine 97%
3,3-Dimethylmorpholine

Precaución: for Research Use Only

Formula: C₆H₁₃NO
MDL Number: MFCD09608072
CAS Number: 59229-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR