4-Bromo-6-ethoxyquinoline 97%

Supplier: Ambeed
Danger

A334347-100MG A334347-1G A334347-250MG
77251-316EA 10 USD
77251-316 77251-318 77251-320
4-Bromo-6-ethoxyquinoline 97%
4-Bromo-6-ethoxyquinoline

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrNO
MW: 252.10 g/mol
MDL Number: MFCD11505111
CAS Number: 1070879-28-5
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR