5-Hydroxy-1,2,3,3-tetramethyl-3H-indol-1-ium iodide 97%

Supplier: Ambeed
Warning

A334175-100MG A334175-1G A334175-250MG
77239-806EA 20 USD
77239-806 77239-808 77239-810
5-Hydroxy-1,2,3,3-tetramethyl-3H-indol-1-ium iodide 97%
5-Hydroxy-1,2,3,3-tetramethyl-3H-indol-1-ium iodide

Precaución: for Research Use Only

Formula: C₁₂H₁₆INO
MDL Number: MFCD28166373
CAS Number: 59223-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR