SAR-125844 98+%

Supplier: Ambeed
Warning

Synonyms: 1-(6-((6-(4-Fluorophenyl)-[1,2,4]triazolo[4,3-b]pyridazin-3-yl)thio)benzo[d]thiazol-2-yl)-3-(2-morpholinoethyl)urea, SAR125844

A334110-10MG A334110-1MG A334110-25MG A334110-5MG
77263-654EA 49.44 USD
77263-654 77263-656 77263-658 77263-660
SAR-125844 98+%
SAR-125844

Precaución: for Research Use Only

Formula: C₂₅H₂₃FN₈O₂S₂
MW: 550.63 g/mol
MDL Number: MFCD25976852
CAS Number: 1116743-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR