5-(Phenylsulfonyl)-N-(piperidin-4-yl)-2-(trifluoromethyl)benzenesulfonamide 98+%

Supplier: Ambeed
Warning

A333983-10MG A333983-1MG A333983-5MG
77239-800EA 45 USD
77239-800 77239-802 77239-804
5-(Phenylsulfonyl)-N-(piperidin-4-yl)-2-(trifluoromethyl)benzenesulfonamide 98+%
5-(Phenylsulfonyl)-N-(piperidin-4-yl)-2-(trifluoromethyl)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₈H₁₉F₃N₂O₄S₂
MDL Number: MFCD18206894
CAS Number: 915759-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR