trans-Dichlorobis(triethylphosphine)palladium(II) 98%

Supplier: Ambeed
Warning

A333948-100MG A333948-250MG
77251-312EA 38.11 USD
77251-312 77251-314
trans-Dichlorobis(triethylphosphine)palladium(II) 98%
trans-Dichlorobis(triethylphosphine)palladium(II)

Precaución: for Research Use Only

Formula: C₁₂H₃₂Cl₂P₂Pd
MW: 415.66 g/mol
MDL Number: MFCD00191831
CAS Number: 28425-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR