3-Bromo-4-methyl-5-nitrobenzoic acid 97%

Supplier: Ambeed
Warning

A333828-1G A333828-250MG A333828-25G A333828-5G
77277-852EA 22 USD
77277-852 77277-854 77277-856 77277-858
3-Bromo-4-methyl-5-nitrobenzoic acid 97%
3-Bromo-4-methyl-5-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrNO₄
MDL Number: MFCD02041260
CAS Number: 34545-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR