(3aR,8aR)-4,4,8,8-Tetrakis(3,5-diisopropylphenyl)-2,2-dimethyl-6-phenyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepine 98%

Supplier: Ambeed
Warning

A333711-100MG A333711-1G A333711-250MG
77251-306EA 83.43 USD
77251-306 77251-308 77251-310
(3aR,8aR)-4,4,8,8-Tetrakis(3,5-diisopropylphenyl)-2,2-dimethyl-6-phenyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepine 98%
(3aR,8aR)-4,4,8,8-Tetrakis(3,5-diisopropylphenyl)-2,2-dimethyl-6-phenyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepine

Precaución: for Research Use Only

Formula: C₆₁H₈₁O₄P
MW: 909.26 g/mol
MDL Number: MFCD23704799
CAS Number: 1361146-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR