D-Argininamide D-arginyl-D-arginyl-D-arginyl-D-arginyl-D-arginyl-D-alanine 97%

Supplier: Ambeed
Warning

A333458-10MG A333458-1MG
77263-644EA 59.74 USD
77263-644 77263-646
D-Argininamide D-arginyl-D-arginyl-D-arginyl-D-arginyl-D-arginyl-D-alanine 97%
D-Argininamide D-arginyl-D-arginyl-D-arginyl-D-arginyl-D-arginyl-D-alanine

Precaución: for Research Use Only

Formula: C₃₆H₇₅N₂₅O₆
MW: 954.14 g/mol
MDL Number: MFCD04974168
CAS Number: 673202-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR