Tracheloside 97%

Supplier: Ambeed
Warning

Synonyms: (3S,4S)-4-(3,4-Dimethoxybenzyl)-3-hydroxy-3-(3-methoxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)benzyl)dihydrofuran-2(3H)-one

A333342-1MG A333342-5MG
77251-298EA 20.6 USD
77251-298 77251-300
Tracheloside 97%
Tracheloside

Precaución: for Research Use Only

Formula: C₂₇H₃₄O₁₂
MW: 550.55 g/mol
MDL Number: MFCD00870631
CAS Number: 33464-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR