1-(4-Bromothiazol-5-yl)ethanone 97%

Supplier: Ambeed
Danger

A333337-100MG A333337-1G A333337-250MG A333337-50MG
77263-636EA 143 USD
77263-636 77263-638 77263-640 77263-642
1-(4-Bromothiazol-5-yl)ethanone 97%
1-(4-Bromothiazol-5-yl)ethanone

Precaución: for Research Use Only

Formula: C₅H₄BrNOS
MDL Number: MFCD22041604
CAS Number: 1368248-75-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR