2-Methyl-3-butenoic acid 95%

Supplier: Ambeed
Danger

A332937-1G A332937-25G A332937-5G
77277-846EA 8 USD
77277-846 77277-848 77277-850
2-Methyl-3-butenoic acid 95%
2-Methyl-3-butenoic acid

Precaución: for Research Use Only

Formula: C₅H₈O₂
MDL Number: MFCD00042650
CAS Number: 53774-20-2
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR