mono-Fmoc ethylene diamine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (9H-Fluoren-9-yl)methyl (2-aminoethyl)carbamate hydrochloride, N-Fmoc-ethylenediamine hydrochloride, Fmoc-NH(CH2)2NH2·HCl

A332835-10G A332835-1G A332835-25G A332835-5G A332835-250MG
77239-784EA 80 USD
77239-784 77239-786 77239-788 77239-790 77616-466
mono-Fmoc ethylene diamine hydrochloride 95%
N-FMOC-ethylenediamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₁₈N₂O₂·HCl
MW: 318.80 g/mol
MDL Number: MFCD00830741
CAS Number: 391624-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR