4-(1H-Pyrazol-3-yl)pyridine 97%

Supplier: Ambeed
Warning

A332657-250MG A332657-1G
77290-470EA 31 USD
77290-470 77748-657
4-(1H-Pyrazol-3-yl)pyridine 97%
4-(1H-Pyrazol-3-yl)pyridine

Precaución: for Research Use Only

Formula: C₈H₇N₃
MDL Number: MFCD00115150
CAS Number: 17784-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR