(2-Fluorophenoxy)acetic acid 97%

Supplier: Ambeed
Warning

A332134-100G A332134-1G A332134-250MG A332134-25G A332134-5G
77263-614EA 179 USD
77263-614 77263-616 77263-618 77263-620 77263-622
(2-Fluorophenoxy)acetic acid 97%
(2-Fluorophenoxy)acetic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₃
MDL Number: MFCD02223229
CAS Number: 348-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR