Dicarbonylbis(triphenylphosphine)nickel 98%

Supplier: Ambeed
Danger

Synonyms: Bis(triphenylphosphine)nickel dicarbonyl, Bis(triphenylphosphine)dicarbonylnickel

A331773-10G A331773-1G A331773-250MG
77263-604EA 75 USD
77263-604 77263-606 77263-608
Dicarbonylbis(triphenylphosphine)nickel 98%
Dicarbonylbis(triphenylphosphine)nickel

Precaución: for Research Use Only

Formula: C₃₈H₃₀NiO₂P₂
MW: 639.29
MDL Number: MFCD00009591
CAS Number: 13007-90-4
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR