(3S,4R,5S)-3,4,5,6-Tetrahydroxy-2-oxohexanoic acid 95%

Supplier: Ambeed
Warning

A331669-1G A331669-25G A331669-5G
77263-594EA 20.6 USD
77263-594 77263-596 77263-598
(3S,4R,5S)-3,4,5,6-Tetrahydroxy-2-oxohexanoic acid 95%
(3S,4R,5S)-3,4,5,6-Tetrahydroxy-2-oxohexanoic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₇
MW: 194.13 g/mol
MDL Number: MFCD06799011
CAS Number: 526-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR