4-Amino-3-chlorophenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-3-chlorobenzeneboronic acid, pinacol ester, 4-Amino-3-chlorophenylboronic Acid Pinacol Ester

A331582-1G A331582-250MG A331582-5G
77290-458EA 25 USD
77290-458 77290-460 77290-462
4-Amino-3-chlorophenylboronic acid pinacol ester 98%
4-Amino-3-chlorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BClNO₂
MDL Number: MFCD09998359
CAS Number: 721960-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR