4-(Diphenylphosphino)benzoic acid 98%

Supplier: Ambeed
Warning

A331506-10G A331506-1G A331506-5G A331506-25G A331506-100G
77290-452EA 203 USD
77290-452 77290-454 77290-456 77748-652 77748-651
4-(Diphenylphosphino)benzoic acid 98%
4-(Diphenylphosphino)benzoic acid

Precaución: for Research Use Only

Formula: C₁₉H₁₅O₂P
MDL Number: MFCD00407264
CAS Number: 2129-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR