2,4-Diphenyl-1H-pyrrole 97%

Supplier: Ambeed
Warning

A331491-100MG A331491-1G A331491-250MG A331491-5G
77251-290EA 41 USD
77251-290 77251-292 77251-294 77251-296
2,4-Diphenyl-1H-pyrrole 97%
2,4-Diphenyl-1H-pyrrole

Precaución: for Research Use Only

Formula: C₁₆H₁₃N
MDL Number: MFCD00958534
CAS Number: 3274-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR