4-Cyano-3-fluorophenyl-4-n-butylbenzoate 97%

Supplier: Ambeed
Warning

A330958-100G A330958-1G A330958-25G A330958-5G
77290-440EA 114 USD
77290-440 77290-442 77290-444 77290-446
4-Cyano-3-fluorophenyl-4-n-butylbenzoate 97%
4-Cyano-3-fluorophenyl-4-n-butylbenzoate

Precaución: for Research Use Only

Formula: C₁₈H₁₆FNO₂
MDL Number: MFCD11053362
CAS Number: 86776-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR