3,9-Divinyl-2,4,8,10-tetraoxaspiro[5.5]undecane 97%

Supplier: Ambeed
Danger

A330919-1G A330919-25G A330919-5G
77277-824EA 5 USD
77277-824 77277-826 77277-828
3,9-Divinyl-2,4,8,10-tetraoxaspiro[5.5]undecane 97%
3,9-Divinyl-2,4,8,10-tetraoxaspiro[5.5]undecane

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₄
MDL Number: MFCD00006564
CAS Number: 78-19-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR