2-Fluoro-6-iodophenol 97%

Supplier: Ambeed
Warning

A330686-10G A330686-1G A330686-250MG A330686-25G A330686-5G
77263-584EA 133 USD
77263-584 77263-586 77263-588 77263-590 77263-592
2-Fluoro-6-iodophenol 97%
2-Fluoro-6-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄FIO
MDL Number: MFCD07782067
CAS Number: 28177-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR