7-Bromo-3-methyl-3H-imidazo[4,5-c]pyridine 95%

Supplier: Ambeed
Warning

A330142-100MG A330142-1G A330142-250MG
77239-760EA 78.28 USD
77239-760 77239-762 77239-764
7-Bromo-3-methyl-3H-imidazo[4,5-c]pyridine 95%
7-Bromo-3-methyl-3H-imidazo[4,5-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MW: 212.04 g/mol
MDL Number: MFCD28578728
CAS Number: 1784125-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR