3-Nitrobiphenyl 98%

Supplier: Ambeed
Warning

A329997-1G A329997-250MG A329997-25G A329997-5G
77290-410EA 15.45 USD
77290-410 77290-412 77290-414 77290-416
3-Nitrobiphenyl 98%
3-Nitrobiphenyl

Precaución: for Research Use Only

Formula: C₁₂H₉NO₂
MW: 199.21 g/mol
MDL Number: MFCD00007245
CAS Number: 2113-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR