3-Iodopyridine-4-carboxylic acid methyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-iodoisonicotinate

A329803-1G A329803-5G
77277-812EA 51.5 USD
77277-812 77277-814
3-Iodopyridine-4-carboxylic acid methyl ester 95%
3-Iodopyridine-4-carboxylic acid methyl ester

Precaución: for Research Use Only

Formula: C₇H₆INO₂
MW: 263.04 g/mol
MDL Number: MFCD03411715
CAS Number: 188677-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR