(4-Nitrophenyl)-α-D-galactopyranoside 97%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol, 4-Nitrophenyl α-D-galactopyranoside

A329708-250MG A329708-1G
77290-402EA 17.51 USD
77290-402 77748-645
(4-Nitrophenyl)-α-D-galactopyranoside 97%
(4-Nitrophenyl)-α-D-galactopyranoside

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₈
MW: 301.26 g/mol
MDL Number: MFCD00065050
CAS Number: 7493-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR