2,4-Diphenyl-4-methyl-1-pentene 95% (by GC)

Supplier: Ambeed
Danger

Synonyms: 4-Methyl-2,4-diphenyl-1-pentene, (4-Methylpent-1-ene-2,4-diyl)dibenzene

A329672-100G A329672-25G A329672-500G
77251-278EA 41.2 USD
77251-278 77251-280 77251-282
2,4-Diphenyl-4-methyl-1-pentene 95% (by GC)
2,4-Diphenyl-4-methyl-1-pentene

Precaución: for Research Use Only

Formula: C₁₈H₂₀
MW: 236.36 g/mol
MDL Number: MFCD00044027
CAS Number: 6362-80-7
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR