1-Acetyl-4-(4-hydroxyphenyl)piperazine 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-(4-Hydroxyphenyl)piperazin-1-yl)ethanone, 4-(4-Acetyl-1-piperazinyl)phenol

A329289-100G A329289-25G A329289-500G
77277-802EA 38.11 USD
77277-802 77277-804 77277-806
1-Acetyl-4-(4-hydroxyphenyl)piperazine 98%
1-Acetyl-4-(4-hydroxyphenyl)piperazine

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O₂
MW: 220.27 g/mol
MDL Number: MFCD00044905
CAS Number: 67914-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR