5-Bromopyrimidine 98%

Supplier: Ambeed
Warning

A328540-100G A328540-10G A328540-1G A328540-25G A328540-500G A328540-5G
77290-390EA 159.65 USD
77290-390 77290-392 77290-394 77290-396 77290-398 77290-400
5-Bromopyrimidine 98%
5-Bromopyrimidine

Precaución: for Research Use Only

Formula: C₄H₃BrN₂
MW: 158.99 g/mol
MDL Number: MFCD00006117
CAS Number: 4595-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR