2,5-Dimethoxybenzonitrile 98%

Supplier: Ambeed
Warning

A327982-1G A327982-25G A327982-5G
77251-262EA 12.36 USD
77251-262 77251-264 77251-266
2,5-Dimethoxybenzonitrile 98%
2,5-Dimethoxybenzonitrile

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MW: 163.18 g/mol
MDL Number: MFCD00016375
CAS Number: 5312-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR