4-Bromo-5-methylthiophene-2-carbaldehyde 98%
Supplier: Ambeed
|
Warning
|
A327948-100MG
A327948-1G
A327948-250MG
A327948-5G
77290-382EA
30.9
USD
77290-382
77290-384
77290-386
77290-388
4-Bromo-5-methylthiophene-2-carbaldehyde 98%
4-Bromo-5-methylthiophene-2-carbaldehyde
Precaución: for Research Use Only
Formula:
C₆H₅BrOS MW: 205.072 g/mol |
MDL Number:
MFCD01118659 CAS Number: 29421-75-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...