3-Fluoro-L-phenylalanine 98%

Supplier: Ambeed
Warning

A327766-100G A327766-100MG A327766-10G A327766-1G A327766-250MG A327766-25G A327766-500G A327766-5G
77251-242EA 567.53 USD
77251-242 77251-244 77251-246 77251-248 77251-250 77251-252 77251-254 77251-256
3-Fluoro-L-phenylalanine 98%
3-Fluoro-L-phenylalanine

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₂
MW: 183.1796 g/mol
MDL Number: MFCD00066450
CAS Number: 19883-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR