(S)-2-Hydroxy-N-methyl-2-phenylacetamide 97%

Supplier: Ambeed
Warning

A327430-1G A327430-5G
77263-554EA 78 USD
77263-554 77263-556
(S)-2-Hydroxy-N-methyl-2-phenylacetamide 97%
(S)-2-Hydroxy-N-methyl-2-phenylacetamide

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MDL Number: MFCD20484260
CAS Number: 65645-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR