2',3'-Dichloroacetophenone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2,3-Dichlorophenyl)ethanone

A327295-100G A327295-10G A327295-1G A327295-25G A327295-5G
77251-232EA 326.51 USD
77251-232 77251-234 77251-236 77251-238 77251-240
2',3'-Dichloroacetophenone 97%
2',3'-Dichloroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
MDL Number: MFCD00052988
CAS Number: 56041-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR