Naquotinib 98%

Supplier: Ambeed
Warning

A327192-10MG A327192-1MG A327192-25MG A327192-5MG
77263-546EA 147 USD
77263-546 77263-548 77263-550 77263-552
Naquotinib 98%
Naquotinib

Precaución: for Research Use Only

Formula: C₃₀H₄₂N₈O₃
MDL Number: MFCD30496701
CAS Number: 1448232-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR