N-(6-((2-Aminophenyl)amino)-6-oxohexyl)-4-methylbenzamide 98%

Supplier: Ambeed
Warning

A327170-10MG A327170-1MG A327170-5MG
77239-692EA 18 USD
77239-692 77239-694 77239-696
N-(6-((2-Aminophenyl)amino)-6-oxohexyl)-4-methylbenzamide 98%
N-(6-((2-Aminophenyl)amino)-6-oxohexyl)-4-methylbenzamide

Precaución: for Research Use Only

Formula: C₂₀H₂₅N₃O₂
MDL Number: MFCD25976845
CAS Number: 1215493-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR