(2-Aminopyridin-3-yl)(phenyl)methanone 95%

Supplier: Ambeed
Warning

A327122-100MG A327122-1G A327122-250MG
77251-220EA 82.4 USD
77251-220 77251-222 77251-224
(2-Aminopyridin-3-yl)(phenyl)methanone 95%
(2-Aminopyridin-3-yl)(phenyl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂O
MW: 198.22 g/mol
MDL Number: MFCD02258896
CAS Number: 3810-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR