3-Hydroxy-4-(trifluoromethyl)benzonitrile 97%

Supplier: Ambeed
Warning

A327042-1G A327042-5G
77263-542EA 26 USD
77263-542 77263-544
3-Hydroxy-4-(trifluoromethyl)benzonitrile 97%
3-Hydroxy-4-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₄F₃NO
MDL Number: MFCD16999170
CAS Number: 731002-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR