3'-Fluoro-4'-(trifluoromethyl)acetophenone 95%

Supplier: Ambeed
Warning

A326938-1G A326938-5G
77277-800EA 17 USD
77277-800 77616-434
3'-Fluoro-4'-(trifluoromethyl)acetophenone 95%
3'-Fluoro-4'-(trifluoromethyl)acetophenone

Precaución: for Research Use Only

Formula: C₉H₆F₄O
MDL Number: MFCD00236273
CAS Number: 237761-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR