(2,4-Difluorobenzyl)hydrazine hydrochloride 97%

Supplier: Ambeed
Warning

A326916-100MG A326916-250MG
77263-532EA 21 USD
77263-532 77263-534
(2,4-Difluorobenzyl)hydrazine hydrochloride 97%
(2,4-Difluorobenzyl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₉ClF₂N₂
MDL Number: MFCD30067271
CAS Number: 1446360-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR