4,4'-((2-Chloro-5-nitrophenyl)methylene)bis(N,N-dimethylaniline) 98%

Supplier: Ambeed
Warning

Synonyms: AL 082D06

A326912-100MG A326912-10MG A326912-1MG A326912-25MG A326912-5MG A326912-50MG
77239-682EA 203.94 USD
77239-682 77239-684 77239-686 77239-688 77239-690 77746-639
4,4'-((2-Chloro-5-nitrophenyl)methylene)bis(N,N-dimethylaniline) 98%
4,4'-((2-Chloro-5-nitrophenyl)methylene)bis(N,N-dimethylaniline)

Precaución: for Research Use Only

Formula: C₂₃H₂₄ClN₃O₂
MW: 409.91 g/mol
MDL Number: MFCD00100145
CAS Number: 256925-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR