4-Bromo-1-ethyl-1H-pyrazole 97%

Supplier: Ambeed
Warning

A326892-100G A326892-10G A326892-25G A326892-5G A326892-1G
77290-362EA 407 USD
77290-362 77290-364 77290-366 77290-368 77571-932
4-Bromo-1-ethyl-1H-pyrazole 97%
4-Bromo-1-ethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₅H₇BrN₂
MDL Number: MFCD08235252
CAS Number: 71229-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR