Methyl 2-cyano-3-oxobutanoate 95%

Supplier: Ambeed
Warning

A326361-100MG A326361-1G A326361-250MG
77263-522EA 28 USD
77263-522 77263-524 77263-526
Methyl 2-cyano-3-oxobutanoate 95%
Methyl 2-cyano-3-oxobutanoate

Precaución: for Research Use Only

Formula: C₆H₇NO₃
MDL Number: MFCD22689506
CAS Number: 3288-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR