4-Methyl-3-(trifluoromethyl)benzoic acid 98%

Supplier: Ambeed
Warning

A326311-100G A326311-10G A326311-1G A326311-25G A326311-5G
77263-512EA 147.99 USD
77263-512 77263-514 77263-516 77263-518 77263-520
4-Methyl-3-(trifluoromethyl)benzoic acid 98%
4-Methyl-3-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂
MDL Number: MFCD01631592
CAS Number: 261952-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR