R112 98+%

Supplier: Ambeed
Warning

Synonyms: 3,3'-((5-Fluoropyrimidine-2,4-diyl)bis(azanediyl))diphenol

A325780-10MG A325780-1MG A325780-25MG A325780-5MG
77239-660EA 28.84 USD
77239-660 77239-662 77239-664 77239-666
R112 98+%
R112

Precaución: for Research Use Only

Formula: C₁₆H₁₃FN₄O₂
MW: 312.30 g/mol
MDL Number: MFCD12922519
CAS Number: 575474-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR