2-(2-Bromoethoxy)anisole 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Bromoethoxy)-2-methoxybenzene

A325049-1G A325049-5G
77239-658EA 29.87 USD
77239-658 77746-633
2-(2-Bromoethoxy)anisole 97%
2-(2-Bromoethoxy)anisole

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₂
MW: 231.09 g/mol
MDL Number: MFCD00017892
CAS Number: 4463-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR