2'-O-Methyluridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-((2R,3R,4R,5R)-4-Hydroxy-5-(hydroxymethyl)-3-methoxytetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 2′-O-Methyluridine
A324867-100G
A324867-10G
A324867-1G
A324867-25G
A324867-5G
77277-772EA
449.08
USD
77277-772
77277-774
77277-776
77277-778
77277-780
2'-O-Methyluridine 98%
2'-O-Methyluridine
Precaución: for Research Use Only
Formula:
C₁₀H₁₄N₂O₆ MW: 258.23 g/mol |
MDL Number:
MFCD00056054 CAS Number: 2140-76-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...