4-Fluoro-3-nitrobenzyl alcohol 98%

Supplier: Ambeed
Warning

A324762-100G A324762-10G A324762-1G A324762-250MG A324762-5G A324762-25G
77290-338EA 330 USD
77290-338 77290-340 77290-342 77290-344 77748-640 77748-639
4-Fluoro-3-nitrobenzyl alcohol 98%
4-Fluoro-3-nitrobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆FNO₃
MDL Number: MFCD01861396
CAS Number: 20274-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR